Learn More
IKA™ ROTAVISC SBS me-vi HELI Complete with UK Plug
GREENER_CHOICE
The SBS (Steel Ball Bearing, Suspended) rotational viscometers extend a large part of the application spectrum of our IKA ROTAVISC device family.
Brand:  IKA™ 0010007718
Product Code. 17564066
  • € 4330.00 / Each
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Description
  • Instead of a jewel bearing, these devices are equipped with a special ball bearing technology.
  • They are characterized by their high robustness. Predestined for applications with very high sample throughput, frequent spindle changes, operation in harsher environments, or even frequently changing device locations.
  • Whether simple or demanding viscosity measurements - with ROTAVISC you get fast and accurate results.
  • Viscosity measurement in motion
  • The motorized IKA HELISTAND moves the mounted viscometer and its rotating body slowly and uniformly up and down in the sample.
  • Easy to use
  • Intuitive menu navigation awaits you via the generous 4.3'' TFT display. A digital leveling also supports the correct setting of the starting condition. Recurring tasks are facilitated by the simple ramp function.
  • Highest measurement accuracy
  • The accuracy of the ROTAVISC rotary viscometers for Newtonian and non-Newtonian liquids is ±1% of the measuring range. The reproducibility is ± 0.2%.
  • Infinitely variable speed
  • With ROTAVISC you have a viscometer in which you can adjust the speed stepless, but also specify programs or individual speed levels.
  • Viscosity-Measuring range: 100 to 40,000,000 mPas
  • Scope of delivery
    • ROTAVISC SBS me-vi control
    • HELISTAND
    • T-SP-SET
    • SP set-2
    • PT 100.8
    • Guard rail
Specifications
Specifications
Viscometer
TFT
0.002
-100°C to +300°C
0.7187 mNm
100/240 V
100 to 40,000,000 mPa
0.01
0.01 to 200 rpm
1 K
50/60 Hz