Learn More
BUCHI Rotavapor Vapor Duct with Combi-Clip
Vapor duct with Combi-Clip for use with connecting the evaporating flask to the Rotavapor.
Brand:  BUCHI 11062187
 View more versions of this product
Product Code. 17603135
  • € 172.00 / Each
Explore more special offers
Description
Description
  • Combi-Clip is included for fastening the evaporating flask
Specifications
Specifications
Borosilicate Glass 3.3
Glass Assembly V, C, S, E, CR, BY, HP
Combi-Clip
Vapor Duct
Rotavapor R-300
24/40