Learn More
ROR1 Mouse anti-Human, Unconjugated, Clone: 1F5D8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66923-1-IG-150UL
 View more versions of this product
Product Code. 16819225
  • € 410.00 / 150µL
Description
The ROR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ROR1. This antibody recognizes human antigen. The ROR1 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ROR1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
1F5D8
Unconjugated
150 μL
4919