Learn More
Merck Millipore ROProtect C Chlorine Tablets
For sanitizing water purification equipment. ROProtect C Chlorine Tablets provide optimal sanitation and protection of reverse osmosis membranes.
Brand: Merck Millipore ZWCL01F50
Description
For sanitizing water purification equipment. ROProtect C Chlorine Tablets provide optimal sanitation and protection of reverse osmosis membranes.
- White tablets
- Supplied in bottles
Safety and Handling
- ROProtect C
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Self-reaC & Dtive substanC & De or mixture Type C & D
- Specific target organ toxicity Category 3
Hazard Statement
- H242-Heating may cause a fire.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : troclosene sodium, dihydrate, adipic acid