Learn More
Thermo Scientific™ Rollout Drawers
Shop All Thermo Scientific Products
Click to view available options
Type:
Additional SS Drawer
Complete SS Drawer Set
Capacity (English):
11.5 cu. ft.
23.3 cu. ft.
29.2 cu. ft.
78.8 cu. ft.
No. of Drawers:
1
6
7
Unit Size:
1 unit
Each
Pack of 6
Description
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
- Stainless-steel drawers
- Adjustable with full extension
Ordering Information:
- Factory installed; must be specified when ordering the refrigerator
Specifications
Specifications
| Type | Additional SS Drawer |
| Capacity (English) | 29.2 cu. ft. |
| Capacity (Metric) | 826 L |
| Drawers | Stainless Steel |
| No. of Drawers | 1 |
| For Use With (Equipment) | 29.2 cu. ft. (826L) Refrigerators and Freezers |