Learn More
Thermo Scientific™ Rollout Drawers
Product Code. 13493918 Shop All Thermo Scientific Products
Click to view available options
Type:
Additional SS Drawer
Complete SS Drawer Set
Capacity (English):
11.5 cu. ft.
23.3 cu. ft.
29.2 cu. ft.
78.8 cu. ft.
No. of Drawers:
1
6
7
Unit Size:
1 unit
Each
Pack of 6
Product Code. 13493918
Brand: Thermo Scientific™ 6699
Only null left
Add to basket
Add to basket
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire refrigerator shelving.
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
  • Stainless-steel drawers
  • Adjustable with full extension
Ordering Information:
Factory installed; must be specified when ordering the refrigerator
TRUSTED_SUSTAINABILITY
Specifications
Type Complete SS Drawer Set
Capacity (English) 23.3 cu. ft.
Capacity (Metric) 659 L
Drawers Stainless Steel
No. of Drawers 6
For Use With (Equipment) 23.3 cu. ft. (659L) Refrigerators and Freezers