Learn More
Horst™ Rod Sensors
Product Code. p-8009519
Click to view available options
Diameter (Metric):
1mm
3mm
Unit Size:
Each
Product Code. 10448641
Brand: Horst™ 70250
Only null left
Specifications
Diameter (Metric) 3mm
Diameter (Metric) Inner 3 mm
Length (Metric) 300 mm
Material PVC
Length (Metric) Power Cord 300mm
Product Type Rod Sensor
Temperature (Metric) 500°C