Learn More
SP Bel-Art™ Robotic Sterile Pipette Tips
Designed for use with automated liquid handling equipment
Brand: SP Bel-Art™ F37860-2005
This item is not returnable.
View return policy
Description
- Sterile, disposable, filtered pipette tips can be used in robotic systems
- Filtered to prevent contamination of pipette arm
- Conductive tip models detect liquid level when pipetting conductive liquids
- Ensures repeatable volume accuracy
- DNase, RNase, pyrogen free, manufactured in Class 8 clean room
- CE marked
Specifications
50 μL | |
Sterile | |
For automated liquid handling equipment | |
2304 | |
No | |
Tecan Genesis Freedom, Freedom EVO and Miniprep with LiHa |
Filtered | |
CE marked | |
24 racks x 96 tips | |
Robotic pipette tips | |
Single-use | |
Polypropylene |