Learn More
Thermo Scientific™ Ring Sets for Precision™ Steaming Water Baths
Precision Ring Sets are for use with steaming water baths.
€ 131.00
Specifications
For Use With (Equipment) Streaming Baths
Type Ring Set
View More Specs
Products 1
Product Code Brand Diameter (English) Price Quantity & Availability  
Product Code Brand Diameter (English) Price Quantity & Availability  
16620583
View Documents Promotion Details
Thermo Scientific™
3166186
10 in.
€ 131.00
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 21-11-2024
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Precision Ring Sets are for use with steaming water baths.
  • Stainless steel
  • Nest smoothly and become progressively larger as rings are removed
  • Three ring sizes available: 10 cm (4in.), 12.5cm (5in.), and 15cm (6in.)
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Streaming Baths
Ring Set