missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Rigid Polypropylene Prohibition Signs
€ 12.39 - € 40.80
Specifications
Material | Polypropylene |
---|---|
Description | Rigid Polypropylene |
For Use With (Application) | Used for a wide variety of applications both indoors and outdoors |
Width (Metric) | 105 mm |
Shape | Round |
Product Code | Brand | Displayed Text | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Price | Quantity & Availability | ||||||
11725014
|
Brady™
807006 |
Defense to use cell phones | N/A | N/A | N/A | |||||
10030624
|
Brady™
252040 |
Prohibits persons with a pacemaker | N/A | N/A | N/A | |||||
11840762
|
Brady™
252052 |
Standing and Parking Prohibited |
€ 13.60
Each |
In Stock
Log in to see stock. |
||||||
Specifications
Polypropylene | |
Used for a wide variety of applications both indoors and outdoors | |
Round |
Rigid Polypropylene | |
105 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Rigid Polypropylene Prohibition Signs