Learn More
Epredia™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes, White
Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Peach
Pink
Tan
White
Yellow
Quantity:
250/box; 2 boxes
500/Cs.
Unit Size:
Pack of 500
Description
- Hinge design ensures specimen safety during processing and easy removal of cover while embedding
- Base is compatible with metal lids and fits all popular size base molds (metal and disposable plastic)
- Cassettes are conveniently available assembled or unassembled to facilitate usage in a labeling system or with metal lids
- Solvent- and decalcifier-resistant and compatible with microwave techniques
- Different colors allow for easy specimen coding
Specifications
Specifications
| Pore Style | Small Square Pore |
| Quantity | 250/box; 2 boxes |
| Closure Type | Hinged |
| Color | White |
| Product Type | Biopsy Cassette |