Learn More
Epredia™ Neg-50™ Frozen Section Medium
Shop All Epredia Products
Click to view available options
Quantity:
4 oz. per bottle
4 oz. per bottle, 6/cs.
4 oz.per bottle
Color:
Blue
Clear
Five Colors plus Clear
Green
Orange
Pink
Yellow
Unit Size:
Pack of 2
Pack of 6
Description
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
- Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
- Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
Specifications
Specifications
| Color | Five Colors plus Clear |
| Packaging Type | 6/Cs. |
| Quantity | 4 oz. per bottle |