Learn More
Epredia™ HistoGel™ Specimen Processing Gel
Product Code. 12823643 Shop All Epredia Products
Click to view available options
Quantity:
1 kit
12 Pk.
144 Pk.
Unit Size:
Each
Pack of 12
Pack of 144
Product Code. 12823643
Brand: Epredia™ HGSK20501
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Encapsulate and retain even small, friable and viscous specimens during histological processing with HistoGel™ Specimen Processing Gel.
  • Penetrates specimen without allowing specimen to escape from gel
  • Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
  • Eliminates unwanted discoloration around specimens on slides
  • Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Twelve 10 mL HistoGel vials, two Epredia™ Richard-Allan Scientific WonderBlock cooling blocks, dry bath incubator, and disposable pipets (500)
Product Type Specimen Processing Gel
Quantity 1 kit