Learn More
Epredia™ Richard-Allan Scientific™ Buffered Wright-Giemsa Stain
Product Code. 12582726 Shop All Thermo Scientific Products
Click to view available options
Quantity:
32 oz.
Unit Size:
Pack of 12
Product Code. 12582726
Brand: Epredia™ 3121
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A single solution stain that contains the fixative, buffer, and stain for rapid and convenient staining of blood smears.
  • Suitable for use on automated systems.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 32 oz.