Learn More
Epredia™ Parapel™ Paraffin Repellent
Protect against excess buildup of wax on countertops and instrumentation with Epredia™ Parapel™ Paraffin Repellent.
Brand:  Epredia™ 2300
Product Code. 10444910
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Low odor
  • Reduces daily cleaning times
  • Will not leave behind oily residue
  • Will not remove finish from surfaces
Specifications
Specifications
4 oz. pump bottle; 6/CS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers