Learn More
Labconco™ ReVo MSC IV Bar
Product Code. 17815602 Shop All Labconco Corporation Products
Click to view available options
Dimensions (L x W x H) Exterior:
900 x 20 x 20 mm
1200 x 20 x 20 mm
1500 x 20 x 20 mm
1800 x 20 x 20 mm
Unit Size:
1 unit
Product Code. 17815602
missing translation for 'mfr': Labconco™ R3858603
Bar supports intravenous solution bottles and bags. Kit includes IV bar and mounting hardware.
  • IV Bar and mounting hardware compatible with various biosafety cabinets, including Purifier Axiom Biosafety Cabinets, Purifier Cell Logic and Cell Logic+ Biosafety Cabinets, Purifier Logic and Logic+ Biosafety Cabinets, Purifier Logic and PuriCare Procedure Stations and ReVo Microbiological Safety Cabinets.
Specifications
Product Type IV Bag Holder Kit
Dimensions (L x W x H) Exterior 1800 x 20 x 20 mm
Shipping Weight (Metric) 3.0 kg