Learn More
Fisherbrand™ Reverse Osmosis Membrane with Housing
Deionizes tap water and filters out particles for laboratory procedures. Fisher Scientific™ Reverse Osmosis Membrane with Housing is for use with the Accu500 Ultrapure Water System.
€ 445.00 - € 464.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
15449669
|
Fisherbrand™
RAR0015HG |
Accu500 15/30L |
€ 464.00
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
15469669
|
Fisherbrand™
RAR0100HG |
Accu500 10/20L |
€ 445.00
Each |
Estimated Shipment: 02-10-2025
Log in to see stock. |
||||||
Description
- For use with Accu500.
- Two sizes: 10L / 20L or 15L / 30L
- Removes particles, ions, and organics from tap water
Specifications
Reverse Osmosis Membrane | |
Housing |
Product Title
Fisherbrand™ Reverse Osmosis Membrane with Housing