Learn More
RETSCH ZM200 Ultra Centrifugal Mill: Ring Sieves
Stainless-steel sieves for use with ZM200 Ultra Centrifugal Mill
Brand:  RETSCH 03.647.0266
 View more versions of this product
Product Code. 11846013
  • € 769.00 / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Specifications
Ring Sieve with Reinforced Rim
Wear-resistant Stainless Steel
Trapezoid holes
ZM200 ultra centrifugal mil
1.0 mm