Learn More
RETSCH PM 100 Planterary Ball Mill Grinding Balls
For use with Retsch PM 100 and PM 200 Planetary Ball Mills
Brand:  RETSCH 22.455.0010
 View more versions of this product
Product Code. 11301460
  • € 475.00 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Description
  • In assorted diameters and materials
  • Smaller balls offered for colloidal milling
Specifications
Specifications
Grinding Ball
110 mL
0.2 cm
PM 100 and PM 200 planetary ball mill
1.1 lb.
3.71 oz.
0.07 in.
Ideal for colloidal milling
Stainless Steel
0.5 kg