Learn More
Applied Biosystems™ resDNASEQ™ Quantitative E1A DNA Fragment Length Kits
The resDNASEQ Quantitative E1A DNA Fragment Length Kit is a quantitative PCR (qPCR)-based system that measures and quantitates the fragment lengths of residual host-cell DNA by targeting the E1A gene in HEK 293 cell lines.
Brand:  Applied Biosystems™ A51970
 View more versions of this product
Product Code. 18715405
  • € 4685.00 / Each
Explore more special offers
Description
Show More
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ resDNASEQ™ Quantitative E1A DNA Fragment Length Kits > Assay Kit and Sample Preparation
For Research Use Only. Not for use in diagnostic procedures.