Learn More
Fisherbrand™ Replacement Bottle Probe
Monitor ambient temperature with this probe. Fisherbrand™ Replacement Bottle Probes may be placed underwater, in refrigerator or freezer.
Brand: Fisherbrand™ 98769-66
This item is not returnable.
View return policy
Description
- Waterproof solid-state probe
- For sentry thermometer
- Sensor and 10-foot cable may be placed underwater, in refrigerator or freezer
- with 3.048m cable
- Temp. Range: -50 to +70°C
Specifications
Solid-state | |
Sentry Thermometer, no. 4121 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Fisherbrand™ Replacement Bottle Probe > Length: 304.8mm; For use with: 4121 Sentry Thermometer