Learn More
Fisherbrand™ Replacement Battery for Electronic Crimpers and Decappers
For electronic Crimpers and Decappers
Brand:  Fisherbrand™ 00 00 3217
228.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 15522300
  • € 242.00 / Each
Explore more special offers
Description
Description
  • Replacement Battery
  • 6.4V Lithium Ion
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lithium Ion 6.4V Battery