Learn More
Thermo Scientific™ Remel™ Violet Red Bile Glucose Agar (Dehydrated)
Product Code. 13426636
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. 13426636
Brand: Thermo Scientific™ R455302
Remel Violet Red Bile Glucose Agar (Dehydrated) is used for selective and differential isolation of Enterobacteriaceae
Remel Violet Red Bile Glucose Agar (Dehydrated) is used for selective and differential isolation of Enterobacteriaceae
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: USP.
Description Violet Red Bile Glucose Agar
Form Powder
Product Type Agar
Quantity 500 g
Not intended for IVD use.