Learn More
Thermo Scientific™ Remel™ Tryptic Soy Broth
Product Code. p-4522919
Click to view available options
Quantity:
10 x 100 mL
Format:
BB Septum - 100mL
Packaging Type:
10/Pk.
Unit Size:
Pack of 10
Product Code. 17719448
Brand: Thermo Scientific™ R112745
The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
  • Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Description Tryptic Soy Broth
Format BB Septum - 100mL
Product Type Microbiology Media
Packaging Type 10/Pk.
Quantity 10 x 100 mL
REF numbers beginning with R11 are not intended for IVD use.