Learn More
Thermo Scientific™ Remel™ Staphaurex™ Disposable Reaction Card, Reaction Cards; 50 cards/pk
Thermo Scientific™ Remel Staphaurex and Disposable Reaction Card is used with Staphaurex and Staphaurex Plus.
Brand: Thermo Scientific™ R30369001
41.71 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Remel Staphaurex and Disposable Reaction Card is used with Staphaurex and Staphaurex Plus.
Specifications
Staphaurex Disposable Reaction Card | |
Yes | |
Reaction Card for Staphylococcus aureus Test Kit | |
50 Cards/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6222041 | Certificate of Analysis | 25/03/2025 | R30369001 |
6182332 | Certificate of Analysis | 14/03/2025 | R30369001 |
6154649 | Certificate of Analysis | 29/01/2025 | R30369001 |
6129050 | Certificate of Analysis | 13/12/2024 | R30369001 |
6129049 | Certificate of Analysis | 06/11/2024 | R30369001 |