Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Brand: Thermo Scientific™ R30166101
439.69 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Specifications
Agglutinating Sera, Haemophilus influenzae Type B | |
H. influenzae | |
Agglutinating Sera |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6159413 | Certificate of Analysis | 03/01/2025 | R30166101 |
6110167 | Certificate of Analysis | 25/07/2024 | R30166101 |
3800011 | Certificate of Analysis | 03/05/2024 | R30166101 |
3697472 | Certificate of Analysis | 04/01/2024 | R30166101 |
3680349 | Certificate of Analysis | 08/09/2023 | R30166101 |