Learn More
Brady™ Reinforced Polypropylene Warning Sign
Brand:  Brady™ 252200
 View more versions of this product
Product Code. 10694725
  • € 10.95 / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Specifications
ISO 7010
Polypropylene
Reinforced Polypropylene
200 mm
200 mm
Triangular
Warning Sign
PIC 300-TRI 200-B7527
1 Ea.
Variety of applications both indoors and outdoors
0.369 mm
-40°C to 120°C
Warning of Flammable Materials
Black on Yellow