Learn More
Brady™ Reinforced Polypropylene Warning Safety Signs
Product Code. 10386004
Click to view available options
Displayed Text:
CAUTION Hot Surface Pictogram
Corrosive Batteries Warning Pictogram
Danger Zone Warning Pictogram - !
Warning of industrial trucks
Height (Metric):
100 mm
178 mm
200 mm
315 mm
Unit Size:
Each
Product Code. 10386004
Brand: Brady™ 252212
Only null left
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC 306-TRI 200-B7527
Material Polypropylene
Quantity 1 Ea.
Description Reinforced Polypropylene
For Use With (Application) Used for a wide variety of applications both indoors and outdoors
Height (Metric) 200 mm
Thickness 0.369 mm
Width (Metric) 200 mm
Temperature Range (Metric) -40°C to 120°C
Shape Triangular
Displayed Text Warning of industrial trucks
Product Type Warning Safety Sign
Color Black on Yellow