Learn More
BINDER™ Reinforced Inner Chamber
Reinforced inner chamber for climate chambers and cooling incubation chambers.
Brand: BINDER™ 8012-2515
Description
- Reinforced inner chamber for bulky and heavy samples.
- Weight capacity ranged from 100 to 200 kg, depending on model.
Specifications
KB PRO 720 | |
Stainless Steel | |
Chamber Rack | |
200 kg |