Learn More
WTW™ Regeneration Set for WTW™ StirrOx™ G
GREENER_CHOICE
Product Code. 10692812
Click to view available options
For Use With (Equipment):
StirrOx G Oxygen Sensor
Unit Size:
Each
Product Code. 10692812
Brand: WTW™ 202710
Only null left
Specifications
For Use With (Equipment) StirrOx G Oxygen Sensor
Type Regeneration Set
Includes Electrolyte, cleaning solution, 3 interchangeable heads foil diaphragm and grinding