Learn More
Kartell™ Plastilab™ Conical One-Hole Stoppers
Made from red rubber.
Brand:  Kartell™ 0382800
 View more versions of this product
Product Code. 10796532
  • € 31.45 / Pack of 25
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Description
Made from red rubber.
  • Made from red rubber
Specifications
Specifications
6mm
25/Pk.
Red
30 mm
Tapered Stopper
29mm
1
21 mm
Rubber