Learn More
Gilac™ Rectangular Container
Product Code. p-200009571
Click to view available options
Volume (Metric):
12 L
15 L
25 L
35 L
55 L
Color:
Gray
White
Unit Size:
Each
Product Code. 17918621
Brand: Gilac™ G119521
Only null left
Add to basket
Add to basket
Rectangular Container is for sub-zero and ambient temperature storage and transporting raw materials.
  • Rectangular plastic food container.
  • For sub-zero and ambient temperature storage and transporting raw materials.
  • No gaps nor overlaps.
  • Machine washable.
  • Covering lid for increased hygiene, optional extra.
  • Stackable without lid.
  • Compatible with all GILAC 4 and 6-wheel dollies.
Specifications
Product Type Food Container
Material Polyethylene
Color White
Shape Rectangular
Volume (English) 3.2 gal.
Volume (Metric) 12 L