Learn More
R&D Systems™ Recombinant SARS-CoV-2 NSP16/NSP10-His Complex
Brand:  R&D Systems™ 10634-CV-100
483.79 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30000892
  • € 624.00 / 100µg
Explore more special offers
Specifications
YP_009725311.1 (NSP16) and YP_009725306.1 (NSP10)
100 μg
SARS-CoV-2
Unconjugated
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Supplied as a 0.2 μm filtered solution in Tris, NaCl, TCEP and Glycerol.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers