Learn More
R&D Systems™ Recombinant Mouse PDGF R alpha Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1062-PR-050
This item is not returnable.
View return policy
Description
The Recombinant Mouse PDGF R alpha Fc Chimera Protein is derived from NS0. The Recombinant Mouse PDGF R alpha Fc Chimera Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
82.5 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
18595 | |
PDGFR alpha | |
Mouse myeloma cell line,NS0-derived mouse PDGF R alpha protein, (N-terminus) Mouse PDGF R alpha Leu25-Glu524 (Asp65Glu,Gly439Ala,Thr440Ala) Accession &Num; P26618.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
Product Content Correction