missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PARK7 Protein
A cDNA sequence encoding the PARK7 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 4090.00
Specifications
Name | PARK7 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952089
|
enQuireBio™
QP12963-5ug |
5 μg |
€ 137.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942089
|
enQuireBio™
QP12963-20ug |
20 μg |
€ 212.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15932089
|
enQuireBio™
QP12963-1mg |
1 mg |
€ 4090.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PARK7 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
PARK7 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASKRAL VILAKGAEEM ETVIPVDVMR RAGIKVTVAG LAGKDPVQCS RDVMICPDTS LEDAKTQGPY DVVVLPGGNL GAQNLSESPM VKEILKEQES RKGLIAAICA GPTALLAHEV GFGCKVTTHP LAKDKMMNGS HYSYSESRVE KDGLILTSRG PGTSFEFALA IVEALVGKDM ANQVKAPLVL KD | |
Greater than 90.0% as determined by SDS-PAGE. |