missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse MMR/CD206 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
€ 582.00
Specifications
Gene ID (Entrez) | 17533 |
---|---|
Molecular Weight (g/mol) | 156 kDa (monomer) |
Name | MMR/CD206 |
Quantity | 50 μg |
Source | Mouse myeloma cell line,NS0-derived mouse MMR/CD206 protein Leu19-Ala1388,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18176550
|
R&D Systems™
2535-MM-050 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
€ 582.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18107810
|
R&D Systems™
2535-MM-050/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
€ 582.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse MMR/CD206 Protein is derived from NS0. The Recombinant Mouse MMR/CD206 Protein has been validated for the following applications: Binding Activity.Specifications
17533 | |
MMR/CD206 | |
Mouse myeloma cell line,NS0-derived mouse MMR/CD206 protein Leu19-Ala1388,with a C-terminal 6-His tag | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
156 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |