missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CCL12 / MCP-5 Protein

A DNA sequence encoding the Mouse CCL12 / MCP-5, was expressed in the hosts and tags indicated.

€ 267.00 - € 1970.00

Specifications

Accession Number NP_035461.2
Gene ID (Entrez) 20293
Name CCL12 / MCP-5 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15953318
View Documents
enQuireBio™
QP10271-ec-5ug
5 μg N/A
N/A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
15933318
View Documents
enQuireBio™
QP10271-ec-20ug
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15923318
View Documents
enQuireBio™
QP10271-ec-100ug
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15943318
View Documents
enQuireBio™
QP10271-ec-250ug
250 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_035461.2
CCL12 / MCP-5 Protein
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
GPDAVSTPVT CCYNVVKQKI HVRKLKSYRR ITSSQCPREA VIFRTILDKE ICADPKEKWV KNSINHLDKT SQTFILEPSC LG
Greater than 95% as determined by SDS-PAGE and HPLC.
20293
Research Use Only
Ccl12
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 10-50 ng/ml.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse CCL12 / MCP-5 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.