missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDT9 Protein
A cDNA sequence encoding the NUDT9 was constructed and used to recombinantly synthesize the protein.
€ 143.00 - € 4090.00
Specifications
Name | NUDT9 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941919
|
enQuireBio™
QP12908-5ug |
5 μg |
€ 143.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15931919
|
enQuireBio™
QP12908-20ug |
20 μg |
€ 222.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15921919
|
enQuireBio™
QP12908-1mg |
1 mg |
€ 4090.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NUDT9 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUDT9 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MNTNVMSGSN GSKENSHNKA RTSPYPGSKV ERSQVPNEKV GWLVEWQDYK PVEYTAVSVL AGPRWADPQI SESNFSPKFN EKDGHVERKS KNGLYEIENG RPRNPAGRTG LVGRGLLGRW GPNHAADPII TRWKRDSSGN KIMHPVSGKH ILQFVAIKRK DCGEWAIPGG MVDPGEKISA TLKREFGEEA LNSLQKTSAE KREIEEKLHK LFSQDHLVIY KGYVDDPRNT DNAWMETEAV NYHDETGEIM DNLMLEAGDD AGKVKWVDIN DKLKLYASHS QFIKLVAEKR DAHWSEDSEA DCHAL | |
Greater than 95.0% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title