missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human MSPR/Ron Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 1947-MS-050
265.22 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human MSP R/Ron Protein is derived from NS0. The Recombinant Human MSP R/Ron Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
60 kDa (single chain), 29.7 kDa ( alpha subunit), 29.6 kDa (beta subunit) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 | |
Recombinant |
4486 | |
MSPR/Ron | |
Mouse myeloma cell line,NS0-derived human MSP R/Ron protein Glu25-Leu571 (single chain) & Gly310-Leu571 (beta),with a C-terminal 10-His tag; Glu25-Arg309 (alpha) | |
Disulfide-linked heterodimer | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |