missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GCNT1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 7248-GT-020
349.88 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to transfer GlcNAc from UDP-GlcNAc to B1-3 galactosyl-N-acetyl galactosamine. The specific activity is >350 pmol/min/μg, as measured under the described conditions.Specifications
NP_001481 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Brij-35. | |
M.W. (Observed): 50-57 kDa, reducing conditions; M.W. (theoretical): 47 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
GCNT1 | |
Recombinant | |
Glucosaminyl (N-acetyl) Transferase 1/GCNT1 |
Enzyme Activity | |
2650 | |
Glucosaminyl (N-acetyl) Transferase 1/GCNT1 | |
Mouse myeloma cell line, NS0-derived human Glucosaminyl (N-acetyl) Transferase 1/GCNT1 protein Arg33-His428, with a C-terminal 6-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.) |