Learn More
R&D Systems™ Recombinant Human FCRN Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 8639-FC-050
404.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18718617
  • € 536.00 / 50µg
Explore more special offers
Description
Description
Non-covalently associated with endogenous beta 2-Microglobulin
Specifications
Specifications
P55899
2217
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FCGRT, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
FCGRT
Unconjugated
Recombinant
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Bioactivity
Observed M.W.: 30-36kDa (FCRN), 10kDa ( beta 2 Microglobulin), Theoretical M.W.: 31kDa (FCRN), 10kDa ( beta 2 Microglobulin)kDa
Human embryonic kidney cell,HEK293-derived human FCRN protein Ala24-Ser297,with a C-terminal 6-His tag
<0.10 EU per 1μg of the protein by the LAL method.
FCRN/FCGRT
Measured by its binding ability in a functional ELISA. When Recombinant Human FCRN is immobilized at 2μg/mL (100μL/well), the concentration of biotinylated human IgG that produces 50% of the optimal binding response is approximately 2-10μg/mL.
Human
Human embryonic kidney cell, HEK293-derived Ala24-Ser297, with a C-terminal 6-His tag
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers