missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EPOR Protein
A cDNA sequence encoding the EPOR was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10591-2ug
This item is not returnable.
View return policy
Specifications
2057 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its ability to inhibit EPO dependent proliferation assay using TF-1 human erythroleukemic cells. The ED50 for this effect less or equal to 70ng/ml. | |
Human | |
His | |
EPOR protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
EPOR Protein | |
Research Use Only | |
EPOR | |
Recombinant Protein | |
Baculovirus | |
APPPNLPDPK FESKAALLAA RGPEELLCFT ERLEDLVCFW EEAASAGVGP GNYSFSYQLE DEPWKLCRLH QAPTARGAVR FWCSLPTADT SSFVPLELRV TAASGAPRYH RVIHINEVVL LDAPVGLVAR LADESGHVVL RWLPPPETPM TSHIRYEVDV SAGNGAGSVQ RVEILEGRTE CVLSNLRGRT RYTFAVRARM AEPSFGGFWS AWSEPVSLLT PSDLDPHHHH HH | |
Greater than 95.0% as determined by analysis by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction