missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Dkk-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1269-DK-010/CF
300.64 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Dkk-4 Protein is derived from NS0. The Recombinant Human Dkk-4 Protein has been validated for the following applications: Bioactivity.Specifications
Bioactivity | |
27121 | |
Dkk-4 | |
Mouse myeloma cell line,NS0-derived human Dkk-4 protein Leu19-Leu224,with a C-terminal 10-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl. | |
24.4 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. | |
Recombinant |