missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Cystatin D Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity
Brand: R&D Systems™ 1202-PI-010
285.09 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Cystatin D Protein is derived from NS0. The Recombinant Human Cystatin D Protein has been validated for the following applications: Inhibition Activity.Specifications
Lyophilized from a 0.2μm filtered solution in MES and NaCl. | |
15 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
1473 | |
Cystatin D | |
Mouse myeloma cell line,NS0-derived human Cystatin D protein Thr29-Val142,with a C-terminal 10-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |