Learn More
R&D Systems™ Recombinant Human Active SYK Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 4594-KS-010
329.15 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Active SYK Protein is derived from Sf 9 (baculovirus). The Recombinant Human Active SYK Protein has been validated for the following applications: Bioactivity.Specifications
Bioactivity | |
Observed MW: 100kDa | |
This product is stable at ≤ -70° C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles. | |
>70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane |
Supplied in 50mM Tris-HCl (pH 7.5), 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol. | |
Spodoptera frugiperda,Sf 9 (baculovirus)-derived human SYK protein Accession &Num; NM_003177 | |
Unconjugated |