Learn More
R&D Systems™ Recombinant Human Active PYK2/FAK2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 4589-KS-010
329.15 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18136460
  • € 418.00 / 10µg
Explore more special offers
Description
Description
The activity of PYK2 is typically 216-292 nmol/min/mg using a poly (Glu:Tyr, 4:1) synthetic peptide substrate (see Activity Assay Protocol).
Specifications
Specifications
NM_004103
Supplied in 50 mM sodium phosphate (pH 7.0), 300 mM NaCl, 0.2 mM DTT, 150 mM imidazole, 0.1 mM PMSF, and 25% glycerol.
M.W. (Observed): 39 kDa
Spodoptera frugiperda, Sf 9 (baculovirus)-derived human PYK2/FAK2 protein Accession # NM_004103
PTK2B
>70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Bioactivity
2185
10 μg
This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.)
Unconjugated
PYK2/FAK2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers