Learn More
Novus Biologicals™ Recombinant GFP Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
Brand:  Novus Biologicals™ NBC1-22949
Product Code. 18755103
  • € 529.00 / 0.10mg
Explore more special offers
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-238 of Aequorea victoria GFP The Recombinant Jellyfish GFP Protein is derived from E. coli. The Recombinant Jellyfish GFP Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
0.1 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers