Learn More
Thermo Scientific™ Reacti-Vap™ Evaporator PTFE-Coated Needles
Shop All Thermo Scientific ProductsBeskrivning
Use these needles to deliver gas and evaporate samples on the Thermo Scientific™ Reacti-Vap™ Evaporator.
Use Thermo Scientific™ Reacti-Vap™ PTFE-Coated Needles to deliver gas and evaporate samples on Reacti-Vap Evaporators. These blunt-ended, 19-gauge, stainless steel needles reduce cross-contamination and corrosion when evaporating solvents that contain strong acids. The needles were specifically designed for Reacti-Vap Evaporators.
Specifikationer
Specifikationer
| Material | PTFE-coated |
| Description | 9 Pack |
| Needle Gauge | 19 G |
| Length (English) Needle | 6 in. |
| Product Type | Syringe Needle |
| Length (Metric) Needle | 152 mm |
| For Use With (Equipment) | Reacti-Vap Evaporators |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.