Learn More
RDH10 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14644-1-AP-150UL
 View more versions of this product
Product Code. 16893384
  • € 410.00 / 150µL
Description
The RDH10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RDH10. This antibody recognizes human, mouse, rat antigen. The RDH10 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
RDH10
Polyclonal
Unconjugated
150 μL
157506
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence
300 μg/mL
Rabbit
Primary
Human, Rat, Mouse