missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat FGF-acidic (FGF-1) Recombinant Protein, PeproTech®

E. coli Recombinant Protein

€ 82.00 - € 1534.70

Specifications

Accession Number P61149
For Use With (Application) Functional Assay
Gene ID (Entrez) 25317
Molecular Weight (g/mol) 15.9
Name Rat FGF-acidic (FGF-1)
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17824233
View Documents
Gibco™
400-29A-100UG
100 μg
€ 304.00
100µg
Please to purchase this item. Need a web account? Register with us today!
17813923
View Documents
Gibco™
400-29A-10UG
10 μg
€ 82.00
10µg
Please to purchase this item. Need a web account? Register with us today!
30088528
View Documents
Gibco™
400-29A-01M
2 x 500 μg
€ 1518.00
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
17804693
View Documents
Gibco™
400-29A-1MG
2 x 500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17854293
View Documents
Gibco™
400-29A-250UG
250 μg
€ 532.00
250µg
Please to purchase this item. Need a web account? Register with us today!
17854693
View Documents
Gibco™
400-29A-500UG
500 μg
€ 912.00
500µg
Please to purchase this item. Need a web account? Register with us today!
17894633
View Documents
Gibco™
400-29A-50UG
50 μg
€ 199.00
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.
Specifications

Specifications

P61149
25317
Rat FGF-acidic (FGF-1)
-20°C
<1 EU/μg
FGF1
Unconjugated
MFNLPLGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SAGEVYIKGT ETGQYLAMDT EGLLYGSQTP NEECLFLERL EENHYNTYTS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D
≥ 95% by SDS-PAGE gel and HPLC analyses.
Functional Assay
15.9
E. coli
RUO
acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; Heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product
Rat
Recombinant
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.